ENA341296669 Enamine

3-nitro-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11983290
  • Cas Number : 39196-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92533
Availability details for
Quantity
  • Availability
  • Add to Cart