ENA341296671 Enamine

2-(chloromethyl)-5-(2-methoxyphenyl)-1,3-oxazole

  • MDL number MFCD09835628
  • Cas Number : 1094452-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-94262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94262
Availability details for
Quantity
  • Availability
  • Add to Cart