ENA341296673 Enamine

2-(3,5-dichlorophenoxy)acetic acid

  • MDL number MFCD04037337
  • Cas Number : 587-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94265

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94265
Availability details for
Quantity
  • Availability
  • Add to Cart