ENA341296677 Enamine

4-(1,3-benzodioxol-5-yl)-3-ethyl-1H-pyrazol-5-amine

  • MDL number MFCD11105165
  • Cas Number : 1187767-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94272

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94272
Availability details for
Quantity
  • Availability
  • Add to Cart