ENA341296679 Enamine

4,6-dimethyl-2H,3H-[1,2]thiazolo[5,4-b]pyridin-3-one

  • MDL number MFCD00232948
  • Cas Number : 60750-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94274
Availability details for
Quantity
  • Availability
  • Add to Cart