ENA341296684 Enamine

3-[4-nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD04970061
  • Cas Number : 1006459-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92537
Availability details for
Quantity
  • Availability
  • Add to Cart