ENA341296686 Enamine

3-iodo-1-phenyl-1H-pyrazole

  • MDL number MFCD12756563
  • Cas Number : 141998-90-5
  • Molecular Weight : 270.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-iodo-1-phenyl-1H-pyrazole
Inchi Code 1S/C9H7IN2/c10-9-6-7-12(11-9)8-4-2-1-3-5-8/h1-7H
Inchi Key RJCCRXXUBFGPBW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-94285

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-94285
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart