ENA341296691 Enamine

(2,5-difluorophenyl)[3-(methylsulfanyl)phenyl]methanone

  • MDL number MFCD20041385
  • Cas Number : 1332355-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94295
Availability details for
Quantity
  • Availability
  • Add to Cart