ENA341296692 Enamine

4-(2H-1,3-benzodioxol-5-yl)-3-(5-bromofuran-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD13659753
  • Cas Number : 1312440-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94296

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94296
Availability details for
Quantity
  • Availability
  • Add to Cart