ENA341296694 Enamine

2-[(1-ethyl-1H-pyrazol-4-yl)methanesulfonyl]acetic acid

  • MDL number MFCD11215396
  • Cas Number : 1174882-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92538
Availability details for
Quantity
  • Availability
  • Add to Cart