ENA341296704 Enamine

4-(4-hydroxyphenyl)-4-oxobutanoic acid

  • MDL number MFCD06249037
  • Cas Number : 56872-39-0
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94335
Availability details for
Quantity
  • Availability
  • Add to Cart