ENA341296706 Enamine

3-(2,4-difluorophenyl)-1,2-oxazol-5-amine

  • MDL number MFCD11156311
  • Cas Number : 1021245-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94337

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94337
Availability details for
Quantity
  • Availability
  • Add to Cart