ENA341296708 Enamine

4-(2,2-difluoroethanesulfonyl)benzoic acid

  • MDL number MFCD12768592
  • Cas Number : 1178294-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-94339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94339
Availability details for
Quantity
  • Availability
  • Add to Cart