ENA341296710 Enamine

3-(2,2,3,3-tetrafluoropropoxy)benzoic acid

  • MDL number MFCD08696480
  • Cas Number : 937602-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92540
Availability details for
Quantity
  • Availability
  • Add to Cart