ENA341296715 Enamine

2-acetamido-3-(4-hydroxyphenyl)propanoic acid

  • MDL number MFCD00063038
  • Cas Number : 2901-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62...64
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94385

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94385
Availability details for
Quantity
  • Availability
  • Add to Cart