ENA341296716 Enamine

3-{2,4-dimethyl-6-oxo-2H,6H,7H-pyrazolo[3,4-b]pyridin-7-yl}propanoic acid

  • MDL number MFCD09834315
  • Cas Number : 1018165-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92542
Availability details for
Quantity
  • Availability
  • Add to Cart