ENA341296720 Enamine

2-amino-1-(2,6-difluorophenyl)propan-1-ol

  • MDL number MFCD11168235
  • Cas Number : 1038262-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94394

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94394
Availability details for
Quantity
  • Availability
  • Add to Cart