ENA341296914 Enamine

3-amino-2-methyl-3-phenylpropanoic acid hydrochloride

  • MDL number MFCD20502224
  • Cas Number : 180182-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230...232
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94616

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94616
Availability details for
Quantity
  • Availability
  • Add to Cart