ENA341296921 Enamine

4-amino-1-(2-phenylcyclopropyl)pyrrolidin-2-one hydrochloride

  • MDL number MFCD20502231
  • Cas Number : 1354961-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94664
Availability details for
Quantity
  • Availability
  • Add to Cart