ENA341296924 Enamine

(2,4-difluorophenyl)[3-(methylsulfanyl)phenyl]methanamine

  • MDL number MFCD20502234
  • Cas Number : 1354962-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94750

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94750
Availability details for
Quantity
  • Availability
  • Add to Cart