ENA341296925 Enamine

4-(2,5-dimethylpyrrolidin-1-yl)-1H-pyrazole

  • MDL number MFCD20502237
  • Cas Number : 1354958-91-0
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94804
Availability details for
Quantity
  • Availability
  • Add to Cart