ENA341296959 Enamine

3-(5-cyclopropyl-1,2,4-oxadiazol-3-yl)benzoic acid

  • MDL number MFCD12026871
  • Cas Number : 775304-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94634

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94634
Availability details for
Quantity
  • Availability
  • Add to Cart