ENA341296965 Enamine

2-methanesulfonyl-1H-imidazole

  • MDL number MFCD01721366
  • Cas Number : 27098-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-215
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-94642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94642
Availability details for
Quantity
  • Availability
  • Add to Cart