ENA341296966 Enamine

2-{[(oxolan-2-yl)methyl]amino}-1-(pyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD05252557
  • Cas Number : 841248-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-94431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94431
Availability details for
Quantity
  • Availability
  • Add to Cart