ENA341296969 Enamine

2-[(quinazolin-4-yl)amino]ethan-1-ol

  • MDL number MFCD00994941
  • Cas Number : 5423-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-94652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94652
Availability details for
Quantity
  • Availability
  • Add to Cart