ENA341296970 Enamine

1-[2-(2-bromophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-ol

  • MDL number MFCD16112750
  • Cas Number : 1251342-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-94653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94653
Availability details for
Quantity
  • Availability
  • Add to Cart