ENA341296973 Enamine

2-[(cyclopropylmethyl)(methyl)amino]pyridine-4-carboxylic acid

  • MDL number MFCD12815717
  • Cas Number : 1184801-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94659

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94659
Availability details for
Quantity
  • Availability
  • Add to Cart