ENA341296980 Enamine

[2-chloro-6-(4-methylpiperazin-1-yl)phenyl]methanamine

  • MDL number MFCD11191625
  • Cas Number : 1037161-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-94432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94432
Availability details for
Quantity
  • Availability
  • Add to Cart