ENA341296981 Enamine

benzyl N-(1-hydroxy-2-methylpropan-2-yl)carbamate

  • MDL number MFCD12185083
  • Cas Number : 62471-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-148
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94674
Availability details for
Quantity
  • Availability
  • Add to Cart