ENA341296986 Enamine

3-[(quinazolin-4-yl)amino]propan-1-ol

  • MDL number MFCD03011503
  • Cas Number : 446829-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-94718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94718
Availability details for
Quantity
  • Availability
  • Add to Cart