ENA341296997 Enamine

2-(ethylamino)-1-(4-methylphenyl)ethan-1-ol

  • MDL number MFCD11916068
  • Cas Number : 873987-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94742
Availability details for
Quantity
  • Availability
  • Add to Cart