ENA341296998 Enamine

2-amino-5-ethyl-4,5-dihydro-1,3-thiazol-4-one

  • MDL number MFCD00644055
  • Cas Number : 1762-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94743

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94743
Availability details for
Quantity
  • Availability
  • Add to Cart