ENA341296999 Enamine

(2-chlorophenyl)(4-methoxy-2-methylphenyl)methanamine

  • MDL number MFCD11149166
  • Cas Number : 1021017-56-0
  • Molecular Weight : 261.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-94745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94745
Availability details for
Quantity
  • Availability
  • Add to Cart