ENA341297015 Enamine

2-(ethenesulfonyl)-2-methylpropane

  • MDL number MFCD11109754
  • Cas Number : 18288-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94803
Availability details for
Quantity
  • Availability
  • Add to Cart