ENA341297018 Enamine

3-ethoxyspiro[3.4]octan-1-amine

  • MDL number MFCD20391446
  • Cas Number : 1342130-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94809

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94809
Availability details for
Quantity
  • Availability
  • Add to Cart