ENA341297026 Enamine

1-{[1-ethyl-3-(propan-2-yl)-1H-pyrazol-5-yl]carbonyl}pyrrolidine-2-carboxylic acid

  • MDL number MFCD13668399

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94842

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94842
Availability details for
Quantity
  • Availability
  • Add to Cart