ENA341297031 Enamine

4-(1,3-benzodioxol-5-yl)-3-cyclopropyl-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD12653263
  • Cas Number : 1355609-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94894

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94894
Availability details for
Quantity
  • Availability
  • Add to Cart