ENA341297040 Enamine

2,2,2-trifluoroethyl N-{4H,5H,6H,7H,8H-cyclohepta[d][1,3]thiazol-2-yl}carbamate

  • MDL number MFCD20502241
  • Cas Number : 1333827-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95041
Availability details for
Quantity
  • Availability
  • Add to Cart