ENA341297068 Enamine

6-chloropyrazine-2-carboxamide

  • MDL number MFCD02258395
  • Cas Number : 36070-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94623
Availability details for
Quantity
  • Availability
  • Add to Cart