ENA346974666 Enamine

2,2,2-trifluoroethyl N-(3-phenylmethanesulfonylpropyl)carbamate

  • MDL number MFCD20731090
  • Cas Number : 1354952-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56425
Availability details for
Quantity
  • Availability
  • Add to Cart