ENA346974674 Enamine

1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)-2,2,2-trifluoroethan-1-ol

  • MDL number MFCD20731101
  • Cas Number : 1354951-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90739

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90739
Availability details for
Quantity
  • Availability
  • Add to Cart