ENA346974675 Enamine

2-bromo-4-fluoro-5-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD20731104
  • Cas Number : 1354952-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80...82
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91694

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91694
Availability details for
Quantity
  • Availability
  • Add to Cart