ENA346974682 Enamine

2-amino-1-(2-fluorophenyl)butan-1-ol hydrochloride

  • MDL number MFCD20731113
  • Cas Number : 1354952-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225...227
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94320

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94320
Availability details for
Quantity
  • Availability
  • Add to Cart