ENA346974688 Enamine

2,2,2-trifluoroethyl N-(2-oxo-1,2-dihydropyridin-3-yl)carbamate

  • MDL number MFCD20731121
  • Cas Number : 1354951-83-9
  • Molecular Weight : 236.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94825
Availability details for
Quantity
  • Availability
  • Add to Cart