ENA346974696 Enamine

{3-[(dimethylamino)methyl]phenyl}boronic acid hydrochloride

  • MDL number MFCD20731144
  • Cas Number : 1485417-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95169
Availability details for
Quantity
  • Availability
  • Add to Cart