ENA346974698 Enamine

2-(propan-2-yl)-1,2,3,4-tetrahydroquinoxaline

  • MDL number MFCD20670070
  • Cas Number : 876184-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95241

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95241
Availability details for
Quantity
  • Availability
  • Add to Cart