ENA346974713 Enamine

2-amino-1-(2-chloro-6-fluorophenyl)butan-1-ol hydrochloride

  • MDL number MFCD20731182
  • Cas Number : 1354950-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95620

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95620
Availability details for
Quantity
  • Availability
  • Add to Cart