ENA346974715 Enamine

4-amino-2,4-dimethylpentan-1-ol

  • MDL number MFCD20731184
  • Cas Number : 1354952-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95652
Availability details for
Quantity
  • Availability
  • Add to Cart