ENA346974717 Enamine

4-amino-2-methylhexan-1-ol

  • MDL number MFCD20731186
  • Cas Number : 1354961-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95681

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95681
Availability details for
Quantity
  • Availability
  • Add to Cart