ENA346974719 Enamine

(2R)-4-carbamoyl-2-decanamidobutanoic acid

  • MDL number MFCD20731187
  • Cas Number : 1354940-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95748

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95748
Availability details for
Quantity
  • Availability
  • Add to Cart